missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-PECAM1 Matched Antibody Pair, Abnova
Marke: Abnova H00005175-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-PECAM1 (100μg); Mouse polyclonal anti-PECAM1 (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
ELISA
Spezifikation
| PECAM1 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PECAM1. | |
| CD31/PECAM-1 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PECAM1 (100 μg) 2. Detection antibody: mouse polyclonal anti-PECAM1 (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| PECAM1 | |
| PECAM1 | |
| Affinity Purified | |
| RUO | |
| 5175 | |
| Human | |
| Serum |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur