Learn More
Parainfluenza Virus Type 3 Monoclonal Antibody (F96H), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA17319
Additional Details : Gewicht : 0.01000kg
Beschreibung
The MA1-7319 antibody reacts with Parainfluenza virus 3 in viral and infected tissue samples. MA1-7319 has been successfully used in ELISA and Immunofluorescence applications. The MA1-7319 antibody was raised against Parainfluenza Type 3 antigen.
Parainfluenza Virus Type 3 Monoclonal antibody specifically detects Parainfluenza Virus Type 3 in Virus samples. It is validated for ELISA, Immunocytochemistry.Spezifikation
Parainfluenza Virus Type 3 | |
Monoclonal | |
0.1 mg/mL | |
PBS with BSA and 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA, Immunocytochemistry | |
F96H | |
FITC | |
canine parainfluenza virus; hPIV-3; PIV-3 | |
Parainfluenza Type 3 antigen. | |
100 μg | |
Primary | |
4° C, store in dark | |
Liquid |