Learn More
p70 [S6K] Mouse, Unlabeled, Clone: 16, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 611261
Additional Details : Gewicht : 0.01100kg
Beschreibung
Increased protein synthesis, an obligatory step in cell proliferation, is regulated by the phosphorylation of a number of translational components, such as ribosomal protein S6. p70 S6 Kinase (p70 [S6K]), a cytosolic kinase that is primarily responsible for S6 phosphorylation, belongs to a family of at least six S6 kinases. p70 [S6K] and p85 [S6K] are two isoforms of the same kinase. Although they are encoded by the same gene, p80 [S6K] contains a 23 amino acid N-terminal extension that serves as a nuclear localization sequence. These kinases are located in a p21 ras-independent signaling pathway and are regulated by Ser/Thr phosphorylation in response to mitogenic stimulation. Activation of p70 [S6K] requires sequential phosphorylations within the autoinhibitory domain and at Thr 389. p70 [S6K] associates with PDK-1 and PKC? to form a possible PI3-K signaling complex. The immunosuppressive agent rapamycin forms an intracellular complex with FKBP12 and FRAP (RAFT1) that inhibits phosphorylation of p70 [S6K] at Thr 389 and, in turn, prevents enzyme activation. Thus, p70 [S6K] is an important regulator of cell proliferation and a potential target of agents that modify the immune and proliferative responses. p70 [S6K] has been reported to be observable migrating within a range of 60-70 kD by western blotting.
Host Species: Mouse
Clone: 16
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat p70 [S6K] aa. 2-121
Formula Weight [Chemical]: 60-70kDa
Immunofluorescence, Western Blotting
Spezifikation
p70 [S6K] | |
Monoclonal | |
250μg/mL | |
p70 S6 Kinase | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
16 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Rat p70 [S6K] aa. 2-121 | |
150 μg | |
Cell Biology | |
Human, Mouse, Rat | |
IgG1 |
For Research Use Only.