missing translation for 'onlineSavingsMsg'
Learn More

Oct3/4 (Human Isoform A) Mouse, PerCP-Cy5.5, Clone: O50-808, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 562251

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 15801759

  • 565.00 CHF / 1 Stück
Verfügbar ab: 06-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Development of a multicellular organism from a single fertilized egg is regulated by the coordinated activity of DNA transcription factors. Oct3/4, a member of the POU family of transcription factors, functions in pluripotent cells of early embryonic stem (ES) cell lines and embryonal carcinomas (EC). The human POU5F1 gene can encode various splice variants, two of which are Oct3/4A and Oct3/4B. Both isoforms share identical POU DNA-binding and C-terminal domains but differ in their N-terminal domain. The N-terminal domain of Oct3/4B is inhibitory to the DNA binding domain and therefore cannot stimulate transcription of Oct3/4-dependent genes. Oct3/4B can be detected in both pluripotent and some differentiated cell types in both the nucleus and cytoplasm, but its function is unclear. There is not an equivalent to Oct3/4B in mouse. Oct3/4A is expressed in the nucleus and has been demonstrated to orchestrate the transcription of Oct3/4-dependent genes. It has been demonstrated that the expression of Oct3/4 isoforms can vary greatly in different cell types, and discrimination of these is crucial for assessing Oct3/4 expression and function. The O50-808 monoclonal antibody recognizes human Oct3/4 Isoform A and mouse Oct3/4.

Host Species: Mouse
Clone: O50-808
Isotype: IgG1, κ
Species Reactivity: Human
Immunogen: Human Oct3/4 Isoform A Recombinant Protein

Flow Cytometry

Spezifikation

Spezifikation

Oct3/4 (Human Isoform A)
Monoclonal
PerCP-Cyanine5.5
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human Oct3/4 Isoform A Recombinant Protein
50 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
O50-808
Oct3/4A, Oct-3A, OTF-3, NF-A3, OTF4, POU5F1, MGC22487
Mouse
Affinity Purified
RUO
Human, Mouse
IgG1 κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote