missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYOD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA523078
Additional Details : Gewicht : 0.01000kg
Beschreibung
The target sequence has 90% sequence homology with mouse.
MYOD Polyclonal antibody specifically detects MYOD in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
MYOD | |
Polyclonal | |
Unconjugated | |
MYOD1 | |
AI503393; bHLHc1; Class C basic helix-loop-helix protein 1; CMD1; MYF3; Myf-3; Myoblast determination protein 1; myoblast determination protein 1 homolog; Myod; MYOD1; Myod-1; MYOD1 homolog; myogenic differentiation 1; myogenic factor 1; myogenic factor 3; PUM; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17927, 4654 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1.0 mg/mL | |
PBS with 0.02% sodium azide | |
P10085, P15172 | |
MYOD1 | |
A synthetic peptide made to an N-terminal region of the human MyoD1 protein (within residues 1-150). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |