Learn More
Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 790, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A11357
Additional Details : Gewicht : 0.01000kg
Beschreibung
This secondary antibody is designed for fluorescent Western blot detection on various near-infrared fluorescence instruments. This antibody can be used for multi-color and multiplexing detection when using other antibodies conjugated to compatible Alexa Fluor™ dyes and wavelengths. Other applications of this antibody include immunofluorescent and fluorescent imaging applications when using instrumentation with appropriate excitation and detection capabilities. Product will be shipped at Room Temperature.
Mouse IgG (H+L) Highly Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Highly Cross-Adsorbed in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Spezifikation
Mouse IgG (H+L) Highly Cross-Adsorbed | |
Polyclonal | |
Alexa Fluor 790 | |
Goat | |
purified | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
1 mg | |
Secondary | |
4° C, store in dark | |
IgG |