Learn More
MKP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528961
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431. Predicted reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MKP4 Polyclonal antibody specifically detects MKP4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MKP4 | |
Polyclonal | |
Unconjugated | |
DUSP9 | |
dual specificity phosphatase 9; dual specificity protein phosphatase 9; dual-specificity MAP kinase phosphatase 4; Dusp4; DUSP9; MAP kinase phosphatase 4; mitogen-activated protein kinase phosphatase 4; MKP4; MKP-4; Mpk4; Pyst3; serine/threonine specific protein phosphatase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1852 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q99956 | |
DUSP9 | |
Synthetic peptide corresponding to a region within amino acids 269 and 360 of Human MKP-4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |