Learn More
MEK5 Mouse, Unlabeled, Clone: 21, BD
Mouse Monoclonal Antibody
619.00 CHF
Spezifikation
Antigen | MEK5 |
---|---|
Klon | 21 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15825729
|
BD Biosciences
610957 |
150 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
The mitogen-activated protein (MAP) kinase cascade participates in the Ras signal transduction pathway. The MAP kinase cascade consists of MEK kinase (Raf-1 or B-Raf), MAP/ERK kinase (MEK1 or MEK2), and extracellular-regulated protein kinase (ERK1 or ERK2). Raf becomes activated following its interaction with Ras-GTP (activated Ras). Via phosphorylation reactions, Raf activates MEK which, in turn, activates ERK. MEK5, also known as MAP Kinase 5, is a 50kDa protein that is 40% identical to MEK1 and MEK2. Alternative splicing results in two MEK5 isoforms: α (50kDa) and β (40kDa). MEK5β has been reported to be ubiquitously expressed while MEK5α is expressed primarily in the liver and brain. The N-terminal amino acid sequence of MEK5α resembles sequences found in actin cytoskeletal proteins, indicating that MEK5α may associate with the cytoskeleton. However, the role of MEK5 in the MAP kinase pathway and its substrate remain to be identified.
Immunofluorescence, Western Blotting
Spezifikation
MEK5 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
MAP Kinase-5 |
21 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Human MEK5 aa. 13-188 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.