missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD206 Monoclonal Antibody (685641), PE, Invitrogen™
Mouse Monoclonal Antibody
Marke: Thermo Scientific MA523594
Additional Details : Gewicht : 0.00010kg
Beschreibung
CD206 Monoclonal antibody specifically detects CD206 in Human samples. It is validated for Flow Cytometry.Spezifikation
CD206 | |
Monoclonal | |
PE | |
Mrc1 | |
AW259686; bA541I19.1; CD206; CELE_Y39A3CR.4; CLEC13D; CLEC13DL; C-type lectin domain family 13 member D; C-type lectin domain family 13 member D-like; ddp-1; hMR; Human mannose receptor; macrophage mannose receptor 1; Macrophage mannose receptor 1-like protein 1; mannose receptor, C type 1; mannose receptor, C type 1-like 1; Mitochondrial import inner membrane translocase subunit Tim8; MMR; MR; MRC1; MRC1L1; tim-8; Y39A3CR.4 | |
Mouse | |
Protein A/G | |
RUO | |
4360 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
685641 | |
Buffered saline solution with BSA and 0.09% sodium azide | |
P22897 | |
Mrc1 | |
Mouse myeloma cell line NS0-derived recombinant mouse MMR/CD206 Leu19-Lys1383 (Thr399Ala) and (Leu407Phe) | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |