Learn More
Ly-6G and Ly-6C Rat anti-Mouse, FITC, Clone: RB6-8C5, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 553126
Additional Details : Gewicht : 0.01000kg
Beschreibung
The RB6-8C5 antibody reacts with a common epitope on Ly-6G and Ly-6C, previously known as the myeloid differentiation antigen Gr-1. In the bone marrow, the level of antigen expression is directly correlated with granulocyte differentiation and maturation.
Host Species: Rat
Clone: RB6-8C5
Isotype: IgG2b, κ
Species Reactivity: Mouse
Flow Cytometry
Spezifikation
Ly-6G and Ly-6C | |
Monoclonal | |
0.5mg/mL | |
Ly6c, Lymphocyte antigen 6C2; Lymphocyte antigen 6G, Ly6g, Gr-1 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
RB6-8C5 | |
FITC | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Not Reported | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b κ |
For Research Use Only.