missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KSR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538713
Additional Details : Gewicht : 0.00900kg
Spezifikation
KSR2 | |
Polyclonal | |
Unconjugated | |
KSR2 | |
KSR2 | |
A synthetic peptide derived from the internal region of human KSR2 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6VAB6, Q3UVC0 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283455, 333050 | |
-20°C | |
Liquid |