missing translation for 'onlineSavingsMsg'
Learn More

Klotho Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA542021

Additional Details : Gewicht : 0.00010kg

Artikelnummer. 15565192

  • 602.00 CHF / 100 Mikroliter
Verfügbar ab: 18-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Peptide sequence: GRLAPGIRGS PRLGYLVAHN LLLAHAKVWH LYNTSFRPTQ GGQVSIALSS Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%

Klotho Polyclonal antibody specifically detects Klotho in Human samples. It is validated for Immunohistochemistry, Western Blot.
Spezifikation

Spezifikation

Klotho
Polyclonal
Unconjugated
KL
alpha-kl; HFTC3; Kl; Klotho; Klotho peptide; secreted form of Klotho protein
Rabbit
Affinity Chromatography
RUO
9365
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9UEF7
KL
Synthetic peptide directed towards the middle region of human KL
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote