missing translation for 'onlineSavingsMsg'
Learn More

KIR2DS5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA537172

Additional Details : Gewicht : 0.00910kg

Artikelnummer. 15524281

  • 604.00 CHF / 100 Mikroliter
Verfügbar ab: 02-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This antibody detects endogenous protein at a molecular weight of 34 kDa. Purity is >95% by SDS-PAGE.

KIR2DS5 Polyclonal antibody specifically detects KIR2DS5 in Human, Mouse, Rat samples. It is validated for Western Blot.
Spezifikation

Spezifikation

KIR2DS5
Polyclonal
Unconjugated
KIR2DS5
CD158 antigen-like family member G; CD158g; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 5; killer cell immunoglobulin-like receptor 2DS5; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5; KIR2DS5; MHC class I NK cell receptor; natural killer cell inhibitory receptor; natural killer cell receptor; natural killer-associated transcript 9; NKAT9; NKAT-9
Rabbit
Antigen affinity chromatography
RUO
3810
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 0.05% sodium azide; pH 7.2
Q14953
KIR2DS5
A synthetic peptide corresponding to residues in human CD158G
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote