Learn More
Cytokeratin 6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532466
Additional Details : Gewicht : 0.01000kg
Beschreibung
Heat-mediated antigen retrieval is recommended prior to staining, using a 1mM EDTA buffer, pH 8.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is prostate or skin tissue.
Cytokeratin 6 Polyclonal antibody specifically detects Cytokeratin 6 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
Cytokeratin 6 | |
Polyclonal | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P02538, P04259, P48668 | |
KRT6A, KRT6B, KRT6C | |
Synthetic peptide derived from C-terminus of human keratin 6 protein | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
KRT6C | |
2310016L08Rik; 60-kDa keratin; BB144589; CK6A; CK-6A; CK6B; CK-6B; CK6C; CK-6C; CK6D; CK-6D; CK-6E; cytokeratin 6A; cytokeratin 6B; cytokeratin 6C; cytokeratin 6D; cytokeratin-6A; Cytokeratin-6B; cytokeratin-6C; Cytokeratin-6D; Cytokeratin-6E; Hom s 5; K6A; K6a keratin; K6B; K6C; K6D; K6E; kamp-keratin derived antimicrobial peptide; KDAMP; Ker2; keratin 6A; keratin 6A, , type II; keratin 6A, type II; keratin 6B; keratin 6B, type II; keratin 6C; keratin 6C, type II; keratin 6E; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6b; keratin complex 2, basic, gene 6c; keratin complex 2, gene 6a; keratin K6; keratin K6h; keratin, epidermal type II, K6A; keratin, epidermal, type II, K6B; keratin, type II cytoskeletal 6A; Keratin, type II cytoskeletal 6B; keratin, type II cytoskeletal 6C; keratin-6A; Keratin-6-alpha; Keratin-6B; keratin-6-beta; keratin-6C; keratin-like 1 (a type II keratin sequence); Krt2-6; Krt2-6a; Krt2-6b; Krt2-6c; Krt6; Krt6a; Krt6b; KRT6C; KRT6D; KRT6E; K | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
286887, 3853, 3854 | |
4° C, do not freeze | |
Liquid |