Learn More
KCC4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA521452
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431, H1299, HepG2, Raji. Predicted reactivity: Mouse (92%), Rat (91%), Rabbit (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
KCC4 Polyclonal antibody specifically detects KCC4 in Human samples. It is validated for Western Blot.Spezifikation
KCC4 | |
Polyclonal | |
Unconjugated | |
SLC12A7 | |
AA408796; D13Ertd261e; electroneutral potassium-chloride cotransporter 4; EMBL:AAH86339.1, ECO:0000312; furosemide-sensitive KCl cotransporter 4; Kcc4; K-Cl cotransporter 4; K-Cl cotransporter KCC4; potassium/chloride transporter KCC4; RGD:1359672}; Slc12a7; slc12a7 {ECO:0000312; solute carrier family 12 (potassium/chloride transporter), member 7; solute carrier family 12 (potassium/chloride transporters), member 7; solute carrier family 12 member 7; solute carrier family 12, member 7 | |
Rabbit | |
Protein A | |
RUO | |
10723 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
4.71 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y666 | |
SLC12A7 | |
Recombinant fragment corresponding to a region within amino acids 845 and 1083 of KCC4 (Uniprot ID#Q9Y666) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |