missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human Kappa Light Chain Secondary Antibody, FITC, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marke: Invitrogen MA110388
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody will not cross-react with hamster, goat, guinea pig, rabbit or sheep.
Human Kappa Light Chain Monoclonal antibody specifically detects Human Kappa Light Chain in Human samples. It is validated for Flow Cytometry.Spezifikation
Human Kappa Light Chain | |
Monoclonal | |
FITC | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
A8B5 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
not available | |
100 Tests | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 |