missing translation for 'onlineSavingsMsg'
Learn More

KIAA0090 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA523732

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 13238848

  • 604.00 CHF / 100 Mikrogramm
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This antibody is predicted to react with chicken based on sequence homology.

KIAA0090 Polyclonal antibody specifically detects KIAA0090 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

KIAA0090
Polyclonal
Unconjugated
EMC1
2700016F22Rik; CAVIPMR; Emc1; ER membrane protein complex subunit 1; KIAA0090; novel DUF1620 domain containing protein; PSEC0263; RGD1310427; RP1-43E13.1
Rabbit
Antigen affinity chromatography, Protein A
RUO
23065, 230866
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
Q8C7X2, Q8N766
EMC1
KLH conjugated synthetic peptide between 866-894 amino acids from the C-terminal region of human K0090
400 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote