missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAM2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530788
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: THP-1. Predicted reactivity: Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
JAM2 Polyclonal antibody specifically detects JAM2 in Human samples. It is validated for Western Blot.Spezifikation
JAM2 | |
Polyclonal | |
Unconjugated | |
JAM2 | |
1110002N23Rik; 2410030G21Rik; 2410167M24Rik; AU016127; C21orf43; CD322; Jam2; JAM-2; JAMB; JAM-B; JAM-IT/VE-JAM; Jcam2; junction adhesion molecule 2; junction adhesion molecule B; junction cell adhesion molecule 2; junctional adhesion molecule 2; Junctional adhesion molecule B; PRO245; RP23-253E17.2; UNQ219/PRO245; Vascular endothelial junction-associated molecule; VEJAM; VE-JAM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58494 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P57087 | |
JAM2 | |
Recombinant fragment corresponding to a region within amino acids 94 and 298 of Human JAM-B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |