Learn More
J Chain Monoclonal Antibody (Mc19-9), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA180527
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody does not detect J chain in intact IgM in ELISA Assays due to steric hindrance. In Western Blotting techniques MA1-80527 detects J chain in IgM only under reducing conditions. Mouse anti Human J chain antibody, clone Mc19-9 reacts with human J chain as tested by ELISA using purified human Ig fragments as the solid phase target antigen.
J Chain Monoclonal antibody specifically detects J Chain in Human samples. It is validated for ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Spezifikation
J Chain | |
Monoclonal | |
1 mg/mL | |
TBS with <0.1% sodium azide | |
P01591 | |
JCHAIN | |
Human J Chain. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
Mc19-9 | |
Unconjugated | |
JCHAIN | |
9530090F24Rik; AI323815; IGCJ; Igj; IgJ chain; immunoglobulin J chain; immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides; immunoglobulin joining chain; immunoglobulin joining gene (not J region); J chain; Jch; JCHAIN; Joining chain of multimeric IgA and IgM | |
Mouse | |
Protein A | |
RUO | |
3512 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |