missing translation for 'onlineSavingsMsg'
Learn More

iNOS Mouse anti-Mouse, Unlabeled, Clone: 54, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 610432

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15825459

  • 615.00 CHF / 1 Set
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Nitric oxide synthase (NOS), a cell-type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits cellular signals involved in vasorelaxation, neurotransmission, and cytotoxicity. In macrophages and other cell types, NOS (iNOS or macNOS) activity increases following exposure to cytokines (IFN-γ, TNF-α, and IL-1) and microbial products (lipopolysaccharide (LPS)). iNOS is activated independently of Ca2+/calmodulin and its level of expression is tightly controlled by several transcription factors, including NFκB. Data indicates that TGF-β affects translation of iNOS mRNA and decreases iNOS protein stability. Normally undetectable in brain tissue, iNOS mRNA has been observed in CNS tissues of animals under experimental pathologic conditions. iNOS and nNOS share 51% amino acid homology with the greatest degree of divergence in the calmodulin binding domain. This antibody has been reported to cross-react with nNOS and eNOS.

Immunofluorescence, Immunoprecipitation, Western Blotting

Spezifikation

Spezifikation

iNOS
Monoclonal
250μg/mL
NOS Type II
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
54
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG2a, κ
150 μg
Cell Biology
Mouse
IgG2a κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.