Learn More
IL-6 Monoclonal Antibody (5IL6), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen M620
Additional Details : Gewicht : 0.80000kg
Beschreibung
M620 was successfully to detect IL-6 in Flow application. M620 targets IL-6 in ELISA, and Western blot applications and shows reactivity with human samples. The M620 immunogen is recombinant human IL-6. M620 detects IL-6 which has a predicted molecular weight of approximately 22 kDa. The M620 IL6 antibody (clone 5IL6) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M621B (biotinylated conjugate of clone 7IL6). Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. Antibody M620 (clone 5IL6) and biotinylated antibody M621B (clone 7IL6) have successfully been used in combination with recombinant IL6 protein SIL6 in ELISA applications. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
IL-6 Monoclonal antibody specifically detects IL-6 in Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Western Blot.Spezifikation
IL-6 | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative | |
P05231, P08505 | |
Il6 | |
Recombinant human IL-6. | |
500 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
ELISA, Flow Cytometry, Western Blot | |
5IL6 | |
Unconjugated | |
Il6 | |
B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 | |
Mouse | |
Protein A | |
RUO | |
16193, 3569 | |
-20°C | |
Liquid |