Learn More
IL2RG Monoclonal Antibody (633162), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524286
Additional Details : Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2 R alpha, rhIL-2 R beta, rhIL-15 R alpha, recombinant mouse IL-2 R gamma, or recombinant canine IL-2 R alpha is observed. In Western blots, approximately 80% cross-reactivity with recombinant human IL-2 R beta, 50% cross-reactivity with rhIL-15 R alpha and recombinant mouse IL-2 R alpha, and no cross-reactivity with rhIL-2 R alpha is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL2RG Monoclonal antibody specifically detects IL2RG in Human samples. It is validated for Flow Cytometry, Neutralization.Spezifikation
IL2RG | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P31785 | |
IL2RG | |
Sf 21-derived recombinant human Common gamma Chain/IL-2 gamma Leu23-Asn254 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Neutralization | |
633162 | |
Unconjugated | |
IL2RG | |
[g]c; Ab2-183; CD132; CD132 antigen; CIDX; common cytokine receptor gamma chain; cytokine receptor common subunit gamma; gamma(c); gammaC; gc; IL-2 receptor subunit gamma; IL2R gamma; IL-2R subunit gamma; IL2RG; IL-2RG; IMD4; interleukin 2 receptor subunit gamma; interleukin 2 receptor, gamma; interleukin 2 receptor, gamma (severe combined immunodeficiency); interleukin 2 receptor, gamma chain; Interleukin-2 receptor subunit gamma; p64; SCIDX; SCIDX1 | |
Mouse | |
Protein A/G | |
RUO | |
3561 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |