missing translation for 'onlineSavingsMsg'
Learn More

IL-2 Rat anti-Human, FITC, Clone: MQ1-17H12, BD

Rat Monoclonal Antibody

Marke:  BD Biosciences 561055

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15830729

  • auf Anfrage / 1 Stück

Dieser Artikel kann nicht online bestellt werden, da Preisänderungen vorbehalten sind. Bitte kontaktieren Sie unser Produktsupport-Team unter 056 618 41 11 für weitere Informationen.


Beschreibung

Beschreibung

The MQ1-17H12 monoclonal antibody specifically binds to the multifunctional cytokine, human Interleukin-2 (IL-2). IL-2 is produced by activated T cells and has multiple functions that can affect the growth, proliferation, differentiation and survival of many different target cell types including T cells, B cells, NK cells, monocytes and macrophages. The immunogen used to generate the MQ1-17H12 hybridoma was purified recombinant human IL-2 protein. The MQ1-17H12 antibody reportedly neutralizes the biological activity of human IL-2.

Spezifikation

Spezifikation

IL-2
MQ1-17H12
FITC
Affinity Purified
Human IL-2 Recombinant Protein
25μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
RUO
Primary
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.