missing translation for 'onlineSavingsMsg'
Learn More

IL-17F Mouse anti-Mouse, Alexa Fluor 488, Clone: O79-289, BD

Mouse Monoclonal Antibody

284.00 CHF

Spezifikation

Antigen IL-17F
Klon O79-289
Konzentration 0.2mg/mL
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 1
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15851209
Unterlagen ansehen Aktionsdetails
BD Biosciences
561658
25 μg Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
N/A Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Beschreibung

Beschreibung

The O79-289 monoclonal antibody specifically binds to the inflammatory cytokine protein, Interleukin-17 (IL-17F). IL-17F is a member of the IL-17 family of cytokines. Among IL-17 family members, IL-17F has the highest amino acid sequence homology to IL-17A. IL-17F is produced by activated CD4+ T helper (Th17) cells, CD8+ T (Tc17) cells and γδ T cells. IL-17F can be secreted as homodimers or as heterodimers with IL-17A. IL-17F and IL-17A have overlapping functions such as inducing epithelial cells and fibroblasts to produce proinflammatory cytokines and chemokines including IL-6, GM-CSF, CXCL1, CCL2, and CCL7. These factors attract and activate neutrophils and other cell types that mediate protective responses against pathogenic microbes or pathologic allergic or autoimmune diseases. IL-17 gene knockout studies have shown that IL-17F and IL-17A have independent functions as well. IL-17F and IL-17A exert their biological function by binding to and signaling through IL-17 receptors comprised of the transmembrane receptor subunits, IL-17RA (CD217) and IL-17RC.

Spezifikation

Spezifikation

IL-17F
0.2mg/mL
Monoclonal
Mouse
Mouse
Mouse IL-17F Recombinant Protein
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
O79-289
Flow Cytometry
Alexa Fluor 488
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
Il17f; interleukin 17F; Interleukin-17F
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

For Research Use Only.