missing translation for 'onlineSavingsMsg'
Learn More

IL-17A Mouse anti-Human, Alexa Fluor 647, Clone: SCPL1362, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 560437

Additional Details : Gewicht : 0.02000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15830199

  • 520.00 CHF / 1 Stück


Verfügbar ab
03-06-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Human IL-17A, also known as IL-17, is a proinflammatory cytokine that is encoded by the IL17A gene in chromosome 6. IL-17A is produced as a disulfide-linked homodimer comprised of two mature 136-amino acid polypeptides. It is a member of the IL-17 family of structurally related cytokines, designated IL-17A through IL-17F. Activated memory T cells, especially Th17 cells (specialized IL-17A-producing CD4+ T cells distinct from Th1 and Th2 cells) produce IL-17 and provide protective immunity against pathogens. Activated CD8+ T cells, γδT cells, NK cells and neutrophils can also be activated to produce IL-17A. IL-17A binds to and exerts its biological activity through IL-17 receptors (IL-17R) that are expressed by a variety of target cells including fibroblasts, epithelial and endothelial cells, monocytes/macrophages and mast cells. The ubiquitous IL-17R expression pattern may explain the broad tissue responsiveness to IL-17. IL-17 induces stromal cells to secrete cytokines and chemokines involved in inflammatory and hematopoietic processes. For example, IL-17 induces fibroblasts to produce IL-6, IL-8, G-CSF and express increased surface ICAM-1. The SCPL1362 antibody reacts with human IL-17A.

Host Species: Mouse
Clone: O33-782
Isotype: IgG1, κ
Species Reactivity: Human

Flow Cytometry, Intracellular Staining

Spezifikation

Spezifikation

IL-17A
SCPL1362
IL-17; IL-17A; CTLA8; Cytotoxic T-lymphocyte-associated serine esterase 8
Affinity Purified
IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
100 Tests
Immunology
Primary
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

For Research Use Only.