missing translation for 'onlineSavingsMsg'
Learn More

IL-17A Rat anti-Mouse, Alexa Fluor 488, Clone: TC11-18H10, BD

Rat Monoclonal Antibody

284.00 CHF - 510.00 CHF

Spezifikation

Antigen IL-17A
Klon TC11-18H10
Konzentration 0.2mg/mL
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15820029
Unterlagen ansehen Aktionsdetails
BD Biosciences
560221
25 μg
284.00 CHF
1 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15810029
Unterlagen ansehen Aktionsdetails
BD Biosciences
560220
0.1 mg
510.00 CHF
1 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

The TC11-18H10 antibody reacts with recombinant and natural mouse IL-17A proteins. IL-17A, also known as CTLA-8, is a T cell-derived cytokine that promotes inflammatory responses. Mouse IL-17A is a proinflammatory cytokine that can induce the release of IL-6 by mouse stromal cells. It has been shown to support the growth of hemopoietic progenitors in vitro; it can also stimulate granulopoiesis in vivo. The TC11-18H10 antibody has been reported to neutralize IL-17A activity. Recent studies have shown that IL-17A is produced by a unique subset of Th17 cells that develop along a pathway distinct from the Th1- and Th2- cell differentiation pathways. The mouse IL-17A cDNA was isolated from a cDNA library generated from TCRαβ+CD4-CD8-thymocytes.

Host Species: Rat
Clone: TC11-18H10
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Recombinant Mouse IL-17A Protein

Intracellular Staining

Spezifikation

Spezifikation

IL-17A
0.2mg/mL
Monoclonal
Rat
Immunology
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
TC11-18H10
Flow Cytometry
Alexa Fluor 488
RUO
Mouse
IgG1 κ
Affinity Purified
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

For Research Use Only.