Learn More
IL-13 Mouse anti-Human, Biotin, Clone: B69-2, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 555054
Beschreibung
The B69-2 antibody reacts with human interleukin-13 (IL-13). The immunogen used to generate the B69-2 hybridoma was recombinant human IL-13 protein. This antibody is routinely tested by ELISA Detection. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
ELISA
Spezifikation
| IL-13 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant human IL-13 protein | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| ELISA | |
| B69-2 | |
| Biotin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.