missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-10 Monoclonal Antibody (945A2A5), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen AHC9102
Additional Details : Gewicht : 0.50000kg
Beschreibung
IL-10 Monoclonal antibody specifically detects IL-10 in Human samples. It is validated for Immunohistochemistry.Spezifikation
IL-10 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.5mg/mL sodium azide; pH 7.5 | |
P22301 | |
IL10 | |
Purified recombinant human IL-10. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry | |
945A2A5 | |
Unconjugated | |
IL10 | |
CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF | |
Mouse | |
Protein A | |
RUO | |
3586 | |
4° C | |
Liquid |