Learn More
IL-10 Rat anti-Mouse, APC, Clone: JES5-16E3, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 554468
Additional Details : Gewicht : 0.01000kg
Beschreibung
The JES5-16E3 antibody reacts with mouse interleukin-10 (IL-10). The immunogen used to generate the JES5-16E3 hybridoma was recombinant mouse IL-10. This is a neutralizing antibody.
Host Species: Rat
Clone: JES5-16E3
Isotype: IgG2b
Species Reactivity: Mouse
Immunogen: Recombinant mouse IL-10
Intracellular Staining
Spezifikation
IL-10 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant mouse IL-10 | |
0.1 mg | |
Immunology | |
Mouse | |
IgG2b |
Flow Cytometry | |
JES5-16E3 | |
APC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.