Learn More
IKAP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529112
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, HepG2, GL261, Raw264.7. Predicted reactivity: Mouse (86%), Rat (85%), Rabbit (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IKAP Polyclonal antibody specifically detects IKAP in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
IKAP | |
Polyclonal | |
Unconjugated | |
ELP1 | |
3110040G09Rik; 6030413P05; C78473; DKFZp781H1425; DYS; elongator acetyltransferase complex subunit 1; elongator complex protein 1; Elp1; FD; FLJ12497; IKAP; IkappaB kinase complex-associated protein; IKBKAP; IKI3; IKK complex-associated protein; IKK-complex-associated protein IKAP; inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein; inhibitor of kappa light polypeptide enhancer in B-cells, kinase complex-associated protein; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein; p150; RP11-3J11.4; TOT1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
230233, 8518 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O95163, Q7TT37 | |
ELP1 | |
Recombinant fragment corresponding to a region within amino acids 768 and 1055 of Human IKBKAP | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |