Learn More
anti-IgG2b, κ Mouse, FITC, Clone: 27-35, BD
Marque:  BD Biosciences 555057
Informations supplémentaires : Gewicht : 0.01000kg
Code produit. 15897978
  • 371.00 CHF / 1 Stück
Verfügbar ab: 27-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Ajouter au panier
Description
Description
The mouse IgG2b, κ immunoglobulin isotype control monoclonal antibody 27-35 is specific for the hapten dansyl (5-[dimethylamino] naphthalene-1-sulfonyl). This hapten is not expressed on human cells or human cell lines. The 27-35 immunoglobulin was selected as an isotype control following testing which demonstrated low background staining on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry, Isotype Control
Spécification
Spécification
Flow Cytometry
0.5mg/mL
Mouse
0.1 mg
IgG2b κ
27-35
FITC
Affinity Purified
Undiluted at 4°C. Protect from exposure to light. Do not freeze.
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Product Certifications