Learn More
anti-IgG2b, κ Mouse, FITC, Clone: 27-35, BD
Marque: BD Biosciences 555057
Informations supplémentaires : Gewicht : 0.01000kg
Description
The mouse IgG2b, κ immunoglobulin isotype control monoclonal antibody 27-35 is specific for the hapten dansyl (5-[dimethylamino] naphthalene-1-sulfonyl). This hapten is not expressed on human cells or human cell lines. The 27-35 immunoglobulin was selected as an isotype control following testing which demonstrated low background staining on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry, Isotype Control
Spécification
Flow Cytometry | |
0.5mg/mL | |
Mouse | |
0.1 mg | |
IgG2b κ |
27-35 | |
FITC | |
Affinity Purified | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |