Learn More
anti-IgG2a κ, Rat, APC, Clone: R35-95, Isotype Control, BD
For research applications
356.00 CHF - 650.00 CHF
Spezifikation
Klon | R35-95 |
---|---|
Anwendungen | Flow Cytometry (Intracellular Staining) |
Konjugat | APC |
Wirtsspezies | Rat |
Immunogen | Mouse Pooled Immunoglobulin |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15807848
|
BD Biosciences
554690 |
0.1 mg |
650.00 CHF
1 Stück |
Verfügbar ab: 27-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15806658
|
BD Biosciences
551442 |
50 Tests |
356.00 CHF
1 Stück |
Verfügbar ab: 27-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Rat
Clone: R35-95
Isotype: IgG2a κ
Immunogen: Mouse Pooled Immunoglobulin
Flow Cytometry, Intracellular Staining, Isotype Control
Spezifikation
R35-95 | |
APC | |
Mouse Pooled Immunoglobulin | |
Affinity Purified |
Flow Cytometry (Intracellular Staining) | |
Rat | |
IgG2a κ | |
Undiluted at 4°C |