Learn More
anti-IgG1 κ, Mouse , PE-Cy™7, Clone: MOPC-21, BD
Marke: BD Biosciences 557646
Additional Details : Gewicht : 0.01000kg
Beschreibung
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Flow Cytometry, Intracellular Staining, Isotype Control
Spezifikation
Flow Cytometry, Western Blot | |
PE-Cyanine7 | |
Affinity Purified | |
Undiluted at 4°C |
MOPC-21 | |
Mouse | |
100 Tests | |
IgG1 κ |