missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG1 κ, Isotype Control, Clone: X40, BD
Marke: BD Biosciences 563231
Additional Details : Gewicht : 0.01000kg
Beschreibung
Host Species: Mouse
Clone:X40
IsotypeIgG1, κ
Immunogen:Keyhole limpet hemocyanin (KLH)
Flow Cytometry, Isotype Control
Spezifikation
Flow Cytometry | |
X40 | |
Brilliant Violet 650 | |
Mouse | |
Affinity Purified | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Keyhole limpet hemocyanin (KLH) | |
50 μg | |
Isotype Control (Primary) | |
IgG1 κ |