Learn More
IFN-γ Rat anti-Mouse, FITC, Clone: XMG1.2, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 562019
128.40 CHF gültig bis 2025-12-31
Benutzen Sie den Promotions-Code "25339" um Ihren Promotions-Preis zu erhalten.
Beschreibung
The XMG1.2 monoclonal antibody specifically binds to mouse interferon- γ(IFN- γ) protein. IFN- γis a pleiotropic cytokine, of approximately 15-17 kDa, involved in the regulation of inflammatory and immune responses. It plays an important role in activation, growth, and differentiation of T and B lymphocytes, macrophages, NK cells and other non-hematopoietic cell types. IFN- γproduction is associated with the Th1 cell differentiation. The purified form of this antibody has been reported to be a neutralizing antibody.

Spezifikation
IFN-γ | |
Monoclonal | |
0.5mg/mL | |
IFN-γ; IFN-g; IFN-gamma; Interferon gamma; Type II Interferon | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
XMG1.2 | |
FITC | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Mouse IFN-γ Recombinant Protein | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.