Learn More
anti-ID2 Matched Antibody Pair, Abnova
Marke: Abnova H00003398-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-ID2 (100μg); Mouse polyclonal anti-ID2 (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Beschreibung
The protein encoded by this gene belongs to the inhibitor of DNA binding (ID) family, members of which are transcriptional regulators that contain a helix-loop-helix (HLH) domain but not a basic domain. Members of the ID family inhibit the functions of basic helix-loop-helix transcription factors in a dominant-negative manner by suppressing their heterodimerization partners through the HLH domains. This protein may play a role in negatively regulating cell differentiation. A pseudogene has been identified for this gene.
ELISA
Spezifikation
| ID2 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ID2. | |
| GIG8/ID2A/ID2H/MGC26389/bHLHb26 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-ID2 (100 μg) 2. Detection antibody: mouse polyclonal anti-ID2 (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| ID2 | |
| ID2 | |
| Affinity Purified | |
| RUO | |
| 3398 | |
| Human | |
| Serum |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.