Learn More
CD90 Mouse anti-Human, Brilliant Violet 605, Clone: 5E10, BD
Mouse Monoclonal Antibody
296.82 CHF - 650.40 CHF
Spezifikation
Antigen | CD90 |
---|---|
Klon | 5.00E+10 |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Konjugat | Brilliant Violet 605 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15842149
|
BD Biosciences
562686 |
25 Tests |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
15832149
|
BD Biosciences
562685 |
100 Tests |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
The 5E10 monoclonal antibody specifically binds to human CD90 which is also known as Thy-1. CD90 is a 25-35 kDa glycophosphatidylinositol-anchored membrane glycoprotein of the Ig superfamily that is expressed on 1-4% of human fetal liver cells, cord blood cells, and bone marrow cells. The anti-CD90 antibody binds to a subset of immature CD34+ cells and a distinct subset of mature CD34- cells that are CD3+CD4+. The CD90+CD34+ population is highly enriched for cells capable of long-term culture. The anti-CD90 antibody is useful for enriching high proliferative potential colony-forming cells (HIPP-CFC) that are primative progenitor cells.
This antibody is conjugated to BD Horizon BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.
Spezifikation
CD90 | |
Flow Cytometry | |
Brilliant Violet 605 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
THY1; Thy-1 antigen; Thy-1 membrane glycoprotein |
5.00E+10 | |
Monoclonal | |
Mouse | |
Human | |
Human HEL Cell Line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.