Learn More
HMGB2, Human, Matched Antibody Pair (AP44), Abnova™
To detect and quantify protein level of human
Marke:  Abnova H00003148-AP44.1Set
Artikelnummer. 16182164
  • ONLINE EXCLUSIVE PRICE
    901.00 CHF
    783.30 CHF / 1 Set
    Sparen 117.70 CHF
    13% Rabatt
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Includes: Capture antibody: mouse monoclonal anti-HMGB2 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-HMGB2, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
HMGB2
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HMGB2.
HMG2
Mouse
RUO
3148
Human
Liquid
ELISA
Biotin
HMGB2
HMGB2
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-HMGB2 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-HMGB2, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications