Learn More
HLA-DR Mouse anti-Human, PE-Cy5, Clone: TU36, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 551375
Beschreibung
Reacts with a monomorphic epitope of major histocompatibility Class II HLA-DR antigens expressed on B cells, activated T cells, and antigen presenting cells. Detects the αβ complex and not the isolated chains. This antibody fixes complement. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: TU36
Isotype: IgG2b κ
Species Reactivity: Human
Flow Cytometry
Spezifikation
| HLA-DR | |
| Monoclonal | |
| PE-Cyanine5 | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Baboon, Canine, Cynomolgus, Human, Rabbit, Rhesus Monkey | |
| IgG2b κ |
| Flow Cytometry | |
| TU36 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.