Learn More
HLA-DR Mouse anti-Human, Brilliant Violet 421, Clone: G46-6, BD
Mouse Monoclonal Antibody
263.22 CHF - 593.85 CHF
Spezifikation
| Antigen | HLA-DR |
|---|---|
| Klon | G46-6 |
| Anwendungen | Flow Cytometry |
| Klassifikation | Monoclonal |
| Konjugat | Brilliant Violet 421 |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15842259
|
BD Biosciences
562805 |
25 Tests |
264.00 CHF 263.22 CHF / 1 Stück Sparen 0.78 CHF 0.30% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
15832259
|
BD Biosciences
562804 |
100 Tests |
595.00 CHF 593.85 CHF / 1 Stück Sparen 1.15 CHF 0.19% Rabatt |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Anti++HLA-DR recognizes a human class II major histocompatibility complex (MHC) antigen. The antigen is a transmembrane glycoprotein composed of α- and β-subunits that have molecular weights of 36 and 27 kDa, respectively. Anti++HLA-DR reacts with a nonpolymorphic HLA-DR epitope and does not cross-react with HLA-DQ or HLA-DP molecules.
Host Species: Mouse
Clone: G46-6
Isotype: IgG2a, κ
Species Reactivity: Hu
Flow Cytometry
Spezifikation
| HLA-DR | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| G46-6 | |
| Monoclonal | |
| Mouse | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| IgG2a κ | |
| Affinity Purified | |
| MHC class II antigen; HLA class II histocompatibility antigen |
For Research Use Only.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.