Learn More
HLA-DR Mouse anti-Human, Biotin, Clone: L243 (G46-6), BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 347361
Additional Details : Gewicht : 0.05710kg
Beschreibung
Antin++DR recognizes a human class II major histocompatibility complex (MHC) antigen. The antigen is a transmembrane glycoprotein composed of α- and β-subunits that have molecular weights of 36 and 27kDa, respectively. Antin++DR reacts with a nonpolymorphic HLA-DR epitope and does not cross-react with HLA-DQ or HLA-DP molecules.
Flow Cytometry
Spezifikation
HLA-DR | |
Monoclonal | |
25μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
100 Tests | |
Immunology | |
Human | |
IgG2a κ |
Flow Cytometry | |
L243 (G46-6) | |
Biotin | |
Mouse | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
For Research Use Only.