missing translation for 'onlineSavingsMsg'
Learn More

GRHL1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA531767

Additional Details : Gewicht : 0.01000kg

Artikelnummer. 13456287

  • 606.00 CHF / 100 Mikroliter
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Recommended positive controls: NT2D1, IMR32, MCF-7. Predicted reactivity: Mouse (91%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GRHL1 Polyclonal antibody specifically detects GRHL1 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

GRHL1
Polyclonal
Unconjugated
GRHL1
DNA methyltransferase 3A; Dnmt3a; grainyhead like 1; grainyhead like transcription factor 1; grainyhead-like 1; grainyhead-like 1 (Drosophila); grainyhead-like protein 1 homolog; grainyhead-like transcription factor 1; GRHL1; LBP protein 32; LBP32; LBP-32; Mammalian grainyhead; Mgr; NH32; p61 MGR; p70 MGR; Tcfcp2l2; TFCP2L2; transcription factor CP2-like 2; Transcription factor LBP-32
Rabbit
Antigen affinity chromatography
RUO
29841
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q9NZI5
GRHL1
Recombinant fragment corresponding to a region within amino acids 378 and 618 of Human GRHL1
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote