missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR146 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533636
Additional Details : Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Marmoset (95%).
GPR146 Polyclonal antibody specifically detects GPR146 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
GPR146 | |
Polyclonal | |
Unconjugated | |
GPR146 | |
BC003323; G protein-coupled receptor 146; Gpr146; G-protein coupled receptor PGR8; PGR8; Probable G-protein coupled receptor 146; RGD1560731 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
115330 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q96CH1 | |
GPR146 | |
Synthetic 20 amino acid peptide from 3rd cytoplasmic domain of human GPR146. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |