missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLPH2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518100
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi transmembrane protein. It processes protein synthesized in the rough endoplasmic reticulum and assists in the transport of protein cargo through the Golgi apparatus. The expression of this encoded protein has been observed to be upregulated in response to viral infection.Spezifikation
GOLPH2 | |
Polyclonal | |
Unconjugated | |
GOLM1 | |
2310001L02Rik; AW125446; bA379P1.3; C9orf155; D030064E01Rik; epididymis luminal protein 46; FLJ22634; FLJ23608; Golgi membrane protein 1; Golgi membrane protein GP73; golgi phosphoprotein 2; golgi protein, 73-kD; GOLM1; Golph2; GOLPH2Ê; GP73; HEL46; PSEC0242; PSEC0257; RP11-379P1.3; UNQ686/PRO1326 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
51280 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8NBJ4 | |
GOLM1 | |
Synthetic peptide sequence (NLLDQREKRNHTL) corresponding to the C-terminus amino acids of GOLM1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |