Learn More
GLUT1 Mouse anti-Human, Mouse, Rat, Clone: SPM498, Invitrogen™
Mouse Monoclonal Antibody
Marke: Thermo Scientific MA511315
Additional Details : Gewicht : 0.01000kg
Beschreibung
MA5-11315 targets GLUT-1 in IHC (P) applications and shows reactivity with Human and Rat samples. The MA5-11315 immunogen is a synthetic peptide derived from C-terminal of human GLUT-1.
Glucose is fundamental to the metabolism in mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.Spezifikation
GLUT1 | |
Monoclonal | |
Unconjugated | |
SLC2A1 | |
GLUT-1, glucose transporter type 1, erythrocyte/brain, hepG2 glucose transporter, solute carrier family 2, facilitated glucose transporter member 1 | |
Mouse | |
Protein A/G | |
RUO | |
20525, 24778, 6513 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
SPM498 | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P11166, P17809, P11167 | |
SLC2A1 | |
A synthetic peptide derived from C-terminal of human GLUT-1 | |
500 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |