Learn More
GLP-1 Monoclonal Antibody (10), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen ABS03310B005
Additional Details : Gewicht : 0.01000kg
Beschreibung
ABS 033-10B-005 detects GLP1 from human samples. ABS 033-10B-005 is specific towards the free N-terminus of GLP-1(7-37) and GLP-1(7-36)amide. ABS 033-10B-005 has been successfully used in ELISA procedures. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
GLP-1 Monoclonal antibody specifically detects GLP-1 in Human samples. It is validated for ELISA.Spezifikation
GLP-1 | |
Monoclonal | |
1.05 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P01275 | |
GCG | |
Synthetic GLP-1(7-17). | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
10 | |
Biotin | |
GCG | |
GCG; Glicentin; Glicentin-related polypeptide; GLP1; GLP-1; GLP-1(7-36); GLP-1(7-37); GLP1/2; GLP2; GLP-2; Glu; glucagon; Glucagon like peptide 1; Glucagon-like peptide 1; Glucagon-like peptide 1(7-36); Glucagon-like peptide 1(7-37); Glucagon-like peptide 2; glucagon-like peptide-1; GRPP; Incretin hormone; MGC138331; OXM; OXY; Oxyntomodulin; PPG; preproglucagon | |
Mouse | |
purified | |
RUO | |
2641 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |