missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSF3R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528988
Additional Details : Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: THP-1, K562. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CSF3R Polyclonal antibody specifically detects CSF3R in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
CSF3R | |
Polyclonal | |
Unconjugated | |
CSF3R | |
CD114; CD114 antigen; colony stimulating factor 3 receptor; colony stimulating factor 3 receptor (granulocyte); Csf3r; Csfgr; G-CSF receptor; GCSFR; G-CSFR; G-CSF-R; Granulocyte colony-stimulating factor receptor; SCN7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1441 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.31 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q99062 | |
CSF3R | |
Recombinant fragment corresponding to a region within amino acids 164 and 432 of Human GCSF Receptor | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |