Learn More
Fyn Mouse, Unlabeled, Clone: 25, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 610164
Beschreibung
Stimulation of the T cell antigen receptor (TcR) results in protein tyrosine phosphorylation via non-receptor tyrosine kinases. The src-related p59[fyn] kinase is one such enzyme that associates with the TcR. Ligation of the TcR activates the protein kinase activity of p59[fyn] invarious human T cells. Fyn interacts with the CD3-ζ chains through its N-terminal region. In turn, fyn binds other proteins through its SH2 and SH3 domains. These proteins (p82 and p116) may serve as substrates and/or mediators of fyn activity. A 72kDa fyn-related protein binds to the TcR/CD3 complex in certain T cells. This protein, known as p72[fyn-R], is a variant of the fyn kinase and contains at least four novel Ser/Thr phosphorylation sites. These sites may make p72[fyn-R] less susceptible to the action of phosphatases than p59[fyn]. Like p59[fyn], p72[fyn-R] is constitutively associated with its receptor. The exact relationship between these two proteins is not yet clear. However, p72[fyn-R] immunoprecipitates with various antisera raised to either the N- or C-terminus of fyn. Similar to src and yes, fyn is highly
expressed in brain. The localization of fyn in mouse brain and its interaction with myelin-associated glycoprotein (MAG) suggests that the fyn kinase has a role in the sensory network and in myelination during early stages of CNS formation.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Spezifikation
| Fyn | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 150 μg | |
| Cell Biology | |
| Canine, Human, Rat | |
| IgG2b |
| Western Blot | |
| 25 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.