missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sensing Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533385
Additional Details : Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Bovine (100%) Marmoset, Rat, Bat, Horse, Rabbit (94%).
Calcium Sensing Receptor Polyclonal antibody specifically detects Calcium Sensing Receptor in Bovine, Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
Calcium Sensing Receptor | |
Polyclonal | |
Unconjugated | |
CASR | |
BoPCaR1; Ca sensing receptor; Ca(2+)-sensing receptor; calcium sensing receptor; calcium-sensing receptor; Calcium-sensing receptor (hypocalciuric hypercalcemia 1 severe neonatal hyperparathyroidism); Calcium-sensing receptor (hypocalciuric hypercalcemia 1, severe neonatal hyperparathyroidism); CaR; CaSR; CaSR antibody; cation sensing receptor; EIG8; Extracellular calcium-sensing receptor; FHH; FIH; G protein coupled receptor, family C, group 2, member A; G protein-coupled receptor, family C, group 2, member A; Gprc2a; HHC; HHC1; HYPOC1; LOW QUALITY PROTEIN: extracellular calcium-sensing receptor; NSHPT; parathyroid Ca(2+)-sensing receptor 1; parathyroid cell calcium-sensing receptor; Parathyroid cell calcium-sensing receptor 1; PCaR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
281038, 846 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P35384, P41180 | |
CASR | |
Synthetic 18 amino acid peptide from N-terminal extracellular domain of human CASR. | |
50 μg | |
Primary | |
Bovine, Human | |
Antibody | |
IgG |