missing translation for 'onlineSavingsMsg'
Learn More

anti-FABP4 Matched Antibody Pair, Abnova™

Marke:  Abnova H00002167-AP21.1Set

Additional Details : Gewicht : 0.00010kg

Artikelnummer. 16172014

  • 850.00 CHF / 1 Set
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FABP4 (100μg); Detection antibody: mouse polyclonal anti-FABP4 (40μL); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols

Beschreibung

Beschreibung

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FABP4

Host Species: Rabbit, Mouse
Species Reactivity: Hu

ELISA Pair (Recombinant Protein)

Spezifikation

Spezifikation

FABP4
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FABP4.
A-FABP/aP2
Mouse, Rabbit
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FABP4 (100 μg) 2. Detection antibody: mouse polyclonal anti-FABP4 (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
Unconjugated
FABP4
FABP4
Affinity Purified
RUO
2167
Human
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote